2-(2-Ethoxyethoxy)ethyl acetate ≥99.0%

Supplier: TCI America

D0500-25G D0500-500G
TCD0500-025GEA 41.08 CAD
TCD0500-025G TCD0500-500G
2-(2-Ethoxyethoxy)ethyl acetate ≥99.0%
2-(2-Ethoxyethoxy)ethyl acetate

CAS Number: 112-15-2
MDL Number: MFCD00041928
Molecular Formula: C8H16O4
Molecular Weight: 176.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 107
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Carbitol Acetate, 2-(2-Ethoxyethoxy)ethyl Acetate

Formula: C₈H₁₆O₄
Boiling Pt: 218 °C
Melting Pt: –25 °C
Density: 1.01 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00041928
CAS Number: 112-15-2
EINECS: 203-940-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f34d115f40ae61fdd3fdc8e339de0c1bbd1f086&vid=1f34d115f40ae61fdd3fdc8e339de0c1bbd1f086&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR