2-(2-Butoxyethoxy)ethyl acetate ≥98.0%
Supplier: TCI America
D0499-25ML
D0499-500ML
TCD0499-25MLEA
24.96
CAD
TCD0499-25ML
TCD0499-500ML
2-(2-Butoxyethoxy)ethyl acetate ≥98.0%
2-(2-Butoxyethoxy)ethyl acetate
CAS Number: 124-17-4
MDL Number: MFCD00009458
Molecular Formula: C10H20O4
Molecular Weight: 204.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Melting point (°C): -32
Flash Point (°C): 124
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2-(2-Butoxyethoxy)ethyl Acetate, Butyl Carbitol Acetate
Formula:
C₁₀H₂₀O₄ Boiling Pt: 247 °C Melting Pt: –32 °C Density: 0.98 (20 °C) Flash Pt: 124 °C |
MDL Number:
MFCD00009458 CAS Number: 124-17-4 EINECS: 204-685-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...