Diethylene glycol ≥99.5%

Supplier: TCI America
Warning

D0495-500G D0495-100G
TCD0495-500GEA 36.91 CAD
TCD0495-500G TCD0495-100G
Diethylene glycol ≥99.5%
Diethylene glycol

CAS Number: 111-46-6
Molecular Formula: C4H10O3
Molecular Weight: 106.12
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Melting point (°C): -7
Flash Point (°C): 124
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 2,2'-Oxydiethanol, Bis(2-hydroxyethyl) Ether

Formula: C₄H₁₀O₃
Boiling Pt: 245 °C
Melting Pt: –7 °C
Density: 1.12 (20 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 13,03146
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47fa5190d36dd34147a57881c675663f97d14cc&vid=a47fa5190d36dd34147a57881c675663f97d14cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR