Silver diethyldithiocarbamate 98,0

Supplier: TCI America
Warning

Synonyms: N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver

D0490-5G D0490-25G
TCD0490-005GEA 164.32 CAD
TCD0490-005G TCD0490-025G
Silver diethyldithiocarbamate 98,0
Silver diethyldithiocarbamate

[for As analysis]
CAS Number: 1470-61-7
MDL Number: MFCD00004929
Molecular Formula: C5H11NS2
Molecular Weight: 256.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: AgDDTC, Diethyldithiocarbamic Acid Silver Salt

Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Melting Pt: 176…178.5 °C
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e92e256b5eb38b21bd24055a08e0e16f4e169cf&vid=bef3328062f5aab5bbd0782035bdf2bede2d9562&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR