Diethylamine hydrochloride ≥98.5%

Supplier: TCI America

Synonyms: Diethylammonium chloride

D0468-500G D0468-25G
TCD0468-500GEA 222.78 CAD
TCD0468-500G TCD0468-025G
Diethylamine hydrochloride ≥98.5%
Diethylamine hydrochloride

CAS Number: 660-68-4
MDL Number: MFCD00012499
Molecular Formula: C4H11N
Molecular Weight: 109.60
Purity/Analysis Method: >98.5% (T)
Form: Crystal
Boiling point (°C): 330
Melting point (°C): 231

Formula: C₄H₁₂ClN
Boiling Pt: 330 °C
Melting Pt: 231 °C
Density: 1.0475 g/cm³ (21 °C)
MDL Number: MFCD00012499
CAS Number: 660-68-4
EINECS: 211-541-9
Merck Index: 13,03138
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be70eb5c44e9e91c50b135a3bd500784917127fd&vid=be70eb5c44e9e91c50b135a3bd500784917127fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR