5-(4-(Diethylamino)benzylidene)rhodanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0464-1G D0464-5G
TCD0464-001GEA 36.34 CAD
TCD0464-001G TCD0464-005G
5-(4-(Diethylamino)benzylidene)rhodanine ≥98.0% (by titrimetric analysis)
5-(4-(Diethylamino)benzylidene)rhodanine

CAS Number: 35778-58-6
MDL Number: MFCD00193758
Molecular Formula: C14H16N2OS2
Molecular Weight: 292.42
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Diethylaminobenzalrhodanine

Formula: C₁₄H₁₆N₂OS₂
MDL Number: MFCD04079831
CAS Number: 35778-58-6
EINECS: 252-723-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7561defbcd8349ced732f34baf9acc121369166&vid=c7561defbcd8349ced732f34baf9acc121369166&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR