4-(Diethylamino)benzaldehyde ≥99.0% (by GC, titration analysis)

Supplier: TCI America

D0463-500G D0463-25G D0463-100G
TCD0463-500GEA 522.98 CAD
TCD0463-500G TCD0463-025G TCD0463-100G
4-(Diethylamino)benzaldehyde ≥99.0% (by GC, titration analysis)
4-(Diethylamino)benzaldehyde

CAS Number: 120-21-8
MDL Number: MFCD00003382
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 42
Flash Point (°C): 165
Storage Temperature: 0-10°C

Formula: C₁₁H₁₅NO
Boiling Pt: 174 °C
Melting Pt: 42 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00003382
CAS Number: 120-21-8
EINECS: 204-377-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44d6c4416fa96de24fa02511aa9e253caaa3636f&vid=ba8e0fcc851fd8abe1beb294ee2b0c11ce5b51de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR