N,N-Diethylacetamide ≥99.0%

Supplier: TCI America
Warning

D0461-25ML D0461-500ML
TCD0461-025MLEA 61.15 CAD
TCD0461-025ML TCD0461-500ML
N,N-Diethylacetamide ≥99.0%
N,N-Diethylacetamide

CAS Number: 685-91-6
MDL Number: MFCD00009047
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 78
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: N-Acetyldiethylamine

Formula: C₆H₁₃NO
Boiling Pt: 185 °C
Density: 0.91 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00009047
CAS Number: 685-91-6
EINECS: 211-685-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=300495fa7b2523ed8c9608b4a97184bfdae7b4dd&vid=300495fa7b2523ed8c9608b4a97184bfdae7b4dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR