2,6-Dichlorophenolindophenol sodium salt dihydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,6-Dichloroindophenol, sodium salt dihydrate, Sodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide dihydrate, Tillmans' reagent dihydrate, 2,6-Dichloroindophenol sodium dihydrate, 2,6-Dichlorophenolindophenol sodium salt, 2,6-Dichloroindophenol sodium salt dihydrate, Sodium 2,6-Dichlorobenzenone-indo-phenol
CAS Number: 620-45-1
MDL Number: MFCD00150015
Molecular Formula: C12H7Cl2NO2
Molecular Weight: 290.07
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Product Categories: Bioscience, Redox Research Reagents
Other Product Names / Synonyms: 2,6-Dichlorophenolindophenol Sodium Salt, Sodium 2,6-Dichlorobenzenone-indo-phenol
Formula:
C₁₂H₆Cl₂NNaO₂·2H₂O MW: 326.105 g/mol Melting Pt: > 300 °C |
MDL Number:
MFCD00150015 CAS Number: 620-45-1 EINECS: 210-640-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...