Ethyl-3,5-dichloro-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Ethyl 3,5-dichloro-4-hydroxybenzoate

D0374-5G D0374-25G
TCD0374-5GEA 101.12 CAD
TCD0374-5G TCD0374-025G
Ethyl-3,5-dichloro-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Ethyl 3,5-dichloro-4-hydroxybenzoate

CAS Number: 17302-82-8
MDL Number: MFCD00016420
Molecular Formula: C9H8Cl2O3
Molecular Weight: 235.06
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 112


Other Product Names / Synonyms: 3,5-Dichloro-4-hydroxybenzoic Acid Ethyl Ester

Formula: C₉H₈Cl₂O₃
MW: 235.07 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00016420
CAS Number: 17302-82-8
EINECS: 241-331-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a28f9a0fcc0433c17585fece503d8043d475f9da&vid=a28f9a0fcc0433c17585fece503d8043d475f9da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR