3,5-Dichloro-4-hydroxybenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
D0373-25G
D0373-5G
TCD0373-025GEA
123.24
CAD
TCD0373-025G
TCD0373-5G
3,5-Dichloro-4-hydroxybenzoic acid ≥98.0%
3,5-Dichloro-4-hydroxybenzoic acid
CAS Number: 3336-41-2
MDL Number: MFCD00002550
Molecular Formula: C7H4Cl2O3
Molecular Weight: 207.01
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Formula:
C₇H₄Cl₂O₃ Boiling Pt: 341 °C (1013 hPa) Melting Pt: 268…269 °C |
MDL Number:
MFCD00002550 CAS Number: 3336-41-2 EINECS: 222-071-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...