Bis(4-chlorophenyl) disulfide ≥98.0%
Supplier: TCI America
Synonyms:
Bis(4-chlorophenyl) disulphide, Bis(4-chlorophenyl) disulfide
D0360-5G
D0360-25G
TCD0360-005GEA
102.7
CAD
TCD0360-005G
TCD0360-025G
Bis(4-chlorophenyl) disulfide ≥98.0%
Bis(4-chlorophenyl) disulphide
CAS Number: 1142-19-4
MDL Number: MFCD00013642
Molecular Formula: C12H8Cl2S2
Molecular Weight: 287.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 72
Other Product Names / Synonyms: Bis(4-Chlorophenyl) Disulfide
Formula:
C₁₂H₈Cl₂S₂ MW: 287.23 g/mol Boiling Pt: 180 °C Melting Pt: 72 °C |
MDL Number:
MFCD00013642 CAS Number: 1142-19-4 EINECS: 214-531-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...