2,6-Dichloro-p-benzoquinone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D0344-5G D0344-1G
TCD0344-005GEA 184.86 CAD
TCD0344-005G TCD0344-001G
2,6-Dichloro-p-benzoquinone ≥98.0% (by titrimetric analysis)
2,6-Dichloro-p-benzoquinone

CAS Number: 697-91-6
MDL Number: MFCD00037159
Molecular Formula: C6H2Cl2O2
Molecular Weight: 176.98
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: 2,6-Dichloro-p-quinone

Formula: C₆H₂Cl₂O₂
Melting Pt: 122 °C
MDL Number: MFCD00037159
CAS Number: 697-91-6
EINECS: 211-810-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca7a7dd49b94e8dcb435456b5f0b5caeb700a99a&vid=ca7a7dd49b94e8dcb435456b5f0b5caeb700a99a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR