2,5-Dichloro-p-benzoquinone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,5-Dichloro-1,4-benzoquinone, 2,5-Dichlorobenzo-1,4-quinone

D0343-5G D0343-1G
TCD0343-005GEA 251.22 CAD
TCD0343-005G TCD0343-001G
2,5-Dichloro-p-benzoquinone ≥98.0% (by HPLC, titration analysis)
2,5-Dichloro-p-benzoquinone

CAS Number: 615-93-0
MDL Number: MFCD00041736
Molecular Formula: C6H2Cl2O2
Molecular Weight: 176.98
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 161


Product Categories: Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2,5-Dichloro-p-quinone

Formula: C₆H₂Cl₂O₂
MW: 176.99 g/mol
Melting Pt: 161 °C
MDL Number: MFCD00041736
CAS Number: 615-93-0
EINECS: 210-453-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e917539f5cf93db311b127e1051f80aaefbb6d77&vid=e917539f5cf93db311b127e1051f80aaefbb6d77&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR