3,5-Dichlorobenzoic acid ≥98.0%

Supplier: TCI America
Warning

D0341-25G D0341-500G
TCD0341-025GEA 41.08 CAD
TCD0341-025G TCD0341-500G
3,5-Dichlorobenzoic acid ≥98.0%
3,5-Dichlorobenzoic acid

CAS Number: 51-36-5
MDL Number: MFCD00002494
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 187

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 309
Melting Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=999fc8dc871c56e851bcc913c269fc0458fa0d53&vid=ac3605f0381b7b1803aec39c6e662aaa9a76fa78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR