2,5-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D0338-25G D0338-100G
TCD0338-025GEA 56.88 CAD
TCD0338-025G TCD0338-100G
2,5-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)
2,5-Dichlorobenzoic acid

CAS Number: 50-79-3
MDL Number: MFCD00002416
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 154

Formula: C₇H₄Cl₂O₂
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002416
CAS Number: 50-79-3
EINECS: 200-065-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=562db9628a40c492eb3d1058d6abbe255f8d569a&vid=86ca7b767814ed61b706de9bff791bfc59fac23e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR