2,4-Dichlorobenzoic acid ≥97.0%
Supplier: TCI America
|
Warning
|
D0337-500G
D0337-25G
TCD0337-500GEA
105.86
CAD
TCD0337-500G
TCD0337-025G
2,4-Dichlorobenzoic acid ≥97.0%
2,4-Dichlorobenzoic acid
CAS Number: 50-84-0
MDL Number: MFCD00002414
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 162
Flash Point (°C): 80
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Boiling Pt: 310 Melting Pt: 162 °C Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002414 CAS Number: 50-84-0 EINECS: 200-067-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...