1,8-Dichloro-9,10-anthraquinone ≥95.0%

Supplier: TCI America

Synonyms: 1,8-Dichloroanthraquinone, 1,8-Dichloroanthracene-9,10-dione

D0329-25G D0329-250G
TCD0329-025GEA 77.42 CAD
TCD0329-025G TCD0329-250G
1,8-Dichloro-9,10-anthraquinone ≥95.0%
1,8-Dichloro-9,10-anthraquinone

CAS Number: 82-43-9
MDL Number: MFCD00001191
Molecular Formula: C14H6Cl2O2
Molecular Weight: 277.10
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 202

Formula: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00001191
CAS Number: 82-43-9
EINECS: 201-420-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e064e9da82bb107d9bfdbceb024ea2577f39c10&vid=1e064e9da82bb107d9bfdbceb024ea2577f39c10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR