1,5-Dichloro-9,10-anthraquinone ≥95.0%

Supplier: TCI America

D0328-25G D0328-500G
TCD0328-025GEA 143.78 CAD
TCD0328-025G TCD0328-500G
1,5-Dichloro-9,10-anthraquinone ≥95.0%
1,5-Dichloro-9,10-anthraquinone

CAS Number: 82-46-2
MDL Number: MFCD00001190
Molecular Formula: C14H6Cl2O2
Molecular Weight: 277.10
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 250

Formula: C₁₄H₆Cl₂O₂
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 250 °C
Density: 1.63 g/cm³ (20 °C)
MDL Number: MFCD00001190
CAS Number: 82-46-2
EINECS: 201-424-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96a21e0cbf3e588add4e2767515b4e8331774ab8&vid=96a21e0cbf3e588add4e2767515b4e8331774ab8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR