9,10-Dichloroanthracene ≥96.0%

Supplier: TCI America
Warning

D0327-10G D0327-1G
TCD0327-010GEA 257.54 CAD
TCD0327-010G TCD0327-001G
9,10-Dichloroanthracene ≥96.0%
9,10-Dichloroanthracene

CAS Number: 605-48-1
MDL Number: MFCD00001246
Molecular Formula: C14H8Cl2
Molecular Weight: 247.12
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 210

Formula: C₁₄H₈Cl₂
MW: 247.12 g/mol
Boiling Pt: 366
Melting Pt: 210 °C
Density: 1.525 g/cm³ (20 °C)
MDL Number: MFCD00001246
CAS Number: 605-48-1
EINECS: 210-087-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a47e2e51ea26eeb9f1d8464c3a61ec13d9223372&vid=dddd433cb67c4c65a782e9da476a22cb4f37d7d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR