Dichloroacetic acid (DCA) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: DCADCA Deblock

D0308-500G D0308-25G
TCD0308-500GEA 774.72 CAD
TCD0308-500G TCD0308-25G
Dichloroacetic acid (DCA) ≥98.0% (by GC, titration analysis)
Dichloroacetic acid (DCA)

CAS Number: 79-43-6
MDL Number: MFCD00004223
Molecular Formula: C2H2Cl2O2
Molecular Weight: 128.94
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 193
Flash Point (°C): 170
Freezing point (°C): 13
Specific Gravity (20/20): 1.57

Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 193 °C
Melting Pt: ∼–4 °C
Density: 1.57 (20 °C)
Flash Pt: 170 °C
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 1764
ADR: 8,II
Merck Index: 13,03075
https://ca.vwr.com/stibo/web/std.lang.all/47/71/28974771.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR