1,3-Dibutyl-2-thiourea ≥97.0% (by total nitrogen basis)

Supplier: TCI America

D0301-500G D0301-25G
TCD0301-500GEA 221.2 CAD
TCD0301-500G TCD0301-25G
1,3-Dibutyl-2-thiourea ≥97.0% (by total nitrogen basis)
1,3-Dibutyl-2-thiourea

CAS Number: 109-46-6
MDL Number: MFCD00004926
Molecular Formula: C9H20N2S
Molecular Weight: 188.33
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Boiling point (°C): 128
Melting point (°C): 65

Formula: H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
Boiling Pt: 128 °C
Melting Pt: 65 °C
MDL Number: MFCD00004926
CAS Number: 109-46-6
EINECS: 203-674-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd1a2856f6a88b74c73d8f35e6444823f401254e&vid=dd1a2856f6a88b74c73d8f35e6444823f401254e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR