1,3-Dimethylurea ≥98.0%

Supplier: TCI America

D0289-25G D0289-500G
TCD0289-025GEA 26.53 CAD
TCD0289-025G TCD0289-500G
1,3-Dimethylurea ≥98.0%
1,3-Dimethylurea

CAS Number: 96-31-1
MDL Number: MFCD00008286
Molecular Formula: C3H8N2O
Molecular Weight: 88.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 105
Flash Point (°C): 157


Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction), Materials Chemistry, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 1,3-Dimethylcarbamide

Formula: C₃H₈N₂O
Boiling Pt: 270 °C
Melting Pt: 105 °C
Density: 1.142 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eecb01e113d8297ec5f4793ffa6963d2ccee1c53&vid=eecb01e113d8297ec5f4793ffa6963d2ccee1c53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR