Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D0281-25G D0281-10G
TCD0281-25GEA 579.86 CAD
TCD0281-25G TCD0281-010G
Diphenylboric acid-2-aminoethyl ester (2-APB) ≥98.0% (by HPLC, titration analysis)
Diphenylboric acid 2-aminoethyl ester (2-APB)

CAS Number: 524-95-8
MDL Number: MFCD00014823
Molecular Formula: C14H16BNO
Molecular Weight: 225.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 190


Product Categories: Bioscience, Inositol Trisphosphate Receptor (IP3R) Antagonists
Other Product Names / Synonyms: (2-Aminoethoxy)diphenylborane, Diphenylborinic Acid 2-Aminoethyl Ester

Formula: C₁₄H₁₆BNO
Melting Pt: 190 °C
MDL Number: MFCD00014823
CAS Number: 524-95-8
EINECS: 208-366-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98e64c219629694f77e057368588907697269d2b&vid=98e64c219629694f77e057368588907697269d2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR