Tetramethyl-1,3-cyclobutanedione ≥99.0%

Supplier: TCI America

D0258-25G D0258-5G
TCD0258-025GEA 227.52 CAD
TCD0258-025G TCD0258-5G
Tetramethyl-1,3-cyclobutanedione ≥99.0%
Tetramethyl-1,3-cyclobutanedione

[Precursor to Dimethyl Ketene]
CAS Number: 933-52-8
MDL Number: MFCD00001331
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 114


Product Categories: Synthetic Organic Chemistry, Precursors for Preparation of Highly Reactive Reagents
Other Product Names / Synonyms: Dimethyl Ketene Dimer

Formula: C₈H₁₂O₂
Boiling Pt: 159 °C
Melting Pt: 114 °C
MDL Number: MFCD00001331
CAS Number: 933-52-8
EINECS: 213-269-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4b773139062f343980eb2e50930e6af9d7d4297&vid=9d21592fb9c680852f825e13671c6910e3d5b027&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR