N,N-Dimethylbenzamide ≥99.0%

Supplier: TCI America

D0256-500G D0256-25G D0256-100G
TCD0256-500GEA 395 CAD
TCD0256-500G TCD0256-025G TCD0256-100G
N,N-Dimethylbenzamide ≥99.0%
N,N-Dimethylbenzamide

CAS Number: 611-74-5
MDL Number: MFCD00008320
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 43
Flash Point (°C): 110

Formula: C₉H₁₁NO
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 43 °C
Density: 1.11 g/cm³ (23 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008320
CAS Number: 611-74-5
EINECS: 210-279-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06895b6f194cbd48c71c689c82b5db73445bde55&vid=06895b6f194cbd48c71c689c82b5db73445bde55&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR