1,4-Dihydroxy-9,10-anthraquinone ≥97.0%

Supplier: TCI America

Synonyms: 1,4-Dihydroxyanthracene-9,10-dione, Quinizarin, 1,4-Dihydroxyanthraquinone

D0243-500G D0243-25G D0243-100G
TCD0243-500GEA 1124.96 CAD
TCD0243-500G TCD0243-025G TCD0243-100G
1,4-Dihydroxy-9,10-anthraquinone ≥97.0%
1,4-Dihydroxy-9,10-anthraquinone

CAS Number: 81-64-1
MDL Number: MFCD00001209
Molecular Formula: C14H8O4
Molecular Weight: 240.21
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Boiling point (°C): 450
Melting point (°C): 196
Flash Point (°C): 222


Other Product Names / Synonyms: 1,4-Dihydroxyanthraquinone

Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 465
Melting Pt: 202
Density: 1.278 g/cm³ (20 °C)
Flash Pt: 222 °C
MDL Number: MFCD00001209
CAS Number: 81-64-1
EINECS: 201-368-7
Merck Index: 12,08250
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66c042b85183df52733ffa3b5dd7eb900497637b&vid=66c042b85183df52733ffa3b5dd7eb900497637b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR