N,N-Dibutylaniline ≥98.0%

Supplier: TCI America
Warning

D0222-25ML D0222-500ML D0222-100ML
TCD0222-025MLEA 82.16 CAD
TCD0222-025ML TCD0222-500ML TCD0222-100ML
N,N-Dibutylaniline ≥98.0%
N,N-Dibutylaniline

CAS Number: 613-29-6
MDL Number: MFCD00035799
Molecular Formula: C14H23N
Molecular Weight: 205.35
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 266
Flash Point (°C): 110
Specific Gravity (20/20): 0.91

Formula: (C₆H₅)N(CH₂CH₂CH₂CH₃)₂
MW: 205.34 g/mol
Boiling Pt: 266 °C
Melting Pt: –32 °C
Density: 0.91 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00035799
CAS Number: 613-29-6
EINECS: 210-335-6
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=591854da6ffbbef8635dced8a9b361486cc58b88&vid=6f71ade11a0cae7f4e3d38dc604afd132d8d9f06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR