1,2-Dibromopropane ≥98.0%

Supplier: TCI America

D0203-25G D0203-500G
TCD0203-025GEA 113.76 CAD
TCD0203-025G TCD0203-500G
1,2-Dibromopropane ≥98.0%
1,2-Dibromopropane

CAS Number: 78-75-1
MDL Number: MFCD00000148
Molecular Formula: C3H6Br2
Molecular Weight: 201.89
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Melting point (°C): -55
Specific Gravity (20/20): 1.93


Other Product Names / Synonyms: Propylene Bromide, Propylene Dibromide

Formula: BrH₂CBrCHCH₃
MW: 201.89 g/mol
Boiling Pt: 141 °C
Melting Pt: –55 °C
Density: 1.93 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000148
CAS Number: 78-75-1
EINECS: 201-139-1
UN: 1993
ADR: 3,III
Merck Index: 12,08038
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=064d98bfcd5579f6ec2991f9a608af440110cc03&vid=2aa09bc7302eb25886729dbff5cee203d813aa76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR