1,10-Dibromodecane ≥95.0%

Supplier: TCI America

Synonyms: 1,10-Dibromodecan, Decane, 1,10-dibromo-

D0179-25G D0179-100G
TCD0179-025GEA 49.61 CAD
TCD0179-025G TCD0179-100G
1,10-Dibromodecane ≥95.0%
1,10-Dibromodecane

CAS Number: 4101-68-2
MDL Number: MFCD00000222
Molecular Formula: C10H20Br2
Molecular Weight: 300.08
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 26


Other Product Names / Synonyms: 1,10-Decamethylene Bromide, 1,10-Decamethylene Dibromide

Formula: BrH₂C(CH₂)₈CH₂Br
MW: 300.08 g/mol
Boiling Pt: 298
Melting Pt: 27
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000222
CAS Number: 4101-68-2
EINECS: 223-871-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f40ceaa0dfc8c9f185a4ca8234bcb06560741f12&vid=befef9547d9637514ba68fb1df70f71167af5c8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR