1,4-Dibromobenzene ≥99.0%

Supplier: TCI America

D0170-500G D0170-25G D0170-100G
TCD0170-500GEA 126.4 CAD
TCD0170-500G TCD0170-025G TCD0170-100G
1,4-Dibromobenzene ≥99.0%
1,4-Dibromobenzene

CAS Number: 106-37-6
MDL Number: MFCD00000089
Molecular Formula: C6H4Br2
Molecular Weight: 235.91
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 219
Melting point (°C): 88
Flash Point (°C): 100
Lambda max.: 282 nm (EtOH)

Formula: C₆H₄Br₂
Boiling Pt: 219 °C
Melting Pt: 88 °C
Density: 2.28 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000089
CAS Number: 106-37-6
EINECS: 203-390-2
UN: 3077
ADR: 9,III
Merck Index: 13,03042
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=692562f03597f167d30c60a5d394ae88464e32d5&vid=aa050eea200beb66e15953052fbac82e8a6c138e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR