1,2-Dibromobenzene ≥97.0%

Supplier: TCI America

D0168-100G D0168-25G
TCD0168-100GEA 225.94 CAD
TCD0168-100G TCD0168-025G
1,2-Dibromobenzene ≥97.0%
1,2-Dibromobenzene

CAS Number: 583-53-9
MDL Number: MFCD00000057
Molecular Formula: C6H4Br2
Molecular Weight: 235.91
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 224
Melting point (°C): 6
Flash Point (°C): 91
Specific Gravity (20/20): 1.99

Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224
Melting Pt: 6 °C
Density: 1.99 (20 °C)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
UN: 3082
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27d7528368d46dd6d461a259ef783a0dc178b33d&vid=c024ff90b9aecd36dcf381e81e16bac5cf909f8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR