9,10-Dibromoanthracene ≥98.0%

Supplier: TCI America
Warning

D0166-25G D0166-500G D0166-100G
TCD0166-025GEA 170.64 CAD
TCD0166-025G TCD0166-500G TCD0166-100G
9,10-Dibromoanthracene ≥98.0%
9,10-Dibromoanthracene

CAS Number: 523-27-3
MDL Number: MFCD00001244
Molecular Formula: C14H8Br2
Molecular Weight: 336.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 227

Formula: C₁₄H₈Br₂
Boiling Pt: 427 °C (1013 hPa)
Melting Pt: 227 °C
Density: 1.981 g/cm³ (20 °C)
MDL Number: MFCD00001244
CAS Number: 523-27-3
EINECS: 208-342-4
Merck Index: 13,03041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19c7c2d3d10b03416bc356763976e7c83026acb9&vid=4f747a0e7ca043a470626201a14df611a4019d23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR