2,4-Diamino-6-phenyl-1,3,5-triazine ≥99.0%

Supplier: TCI America

Synonyms: Benzoguanamine, 2,4-Diamino-6-phenyl-s-triazine, 6-Phenyl-1,3,5-triazine-2,4-diyldiamine, 6-Phenyl-1,3,5-triazine-2,4-diamine, 2,6-Diamino-4-phenyl-1,3,5-triazine

D0111-25G D0111-500G
TCD0111-025GEA 33.76 CAD
TCD0111-025G TCD0111-500G
2,4-Diamino-6-phenyl-1,3,5-triazine ≥99.0%
2,4-Diamino-6-phenyl-1,3,5-triazine

CAS Number: 91-76-9
MDL Number: MFCD00023187
Molecular Formula: C9H9N5
Molecular Weight: 187.21
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 228


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,4-Diamino-6-phenyl-1,3,5-triazine

Formula: C₉H₉N₅
MW: 187.2 g/mol
Melting Pt: 228 °C
MDL Number: MFCD00023187
CAS Number: 91-76-9
EINECS: 202-095-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d15b87ad714ea5ed7108e77f2888e939fcb8fd&vid=f2d15b87ad714ea5ed7108e77f2888e939fcb8fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR