2-Nitro-p-phenylenediamine ≥94.0%

Supplier: TCI America

Synonyms: 2-Nitro-1,4-phenylenediamine, 2-Nitrobenzene-1,4-diamine

D0105-25G D0105-500G
TCD0105-025GEA 47.4 CAD
TCD0105-025G TCD0105-500G
2-Nitro-p-phenylenediamine ≥94.0%
2-Nitro-p-phenylenediamine

CAS Number: 5307-14-2
MDL Number: MFCD00007903
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >94.0% (T)
Form: Crystal
Melting point (°C): 138


Other Product Names / Synonyms: 1,4-Diamino-2-nitrobenzene

Formula: C₆H₇N₃O₂
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007903
CAS Number: 5307-14-2
EINECS: 226-164-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b89134db7531aec8b10f3461bdfdf47e417da4a&vid=2b89134db7531aec8b10f3461bdfdf47e417da4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR