4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D0103-25G
D0103-100G
TCD0103-025GEA
40.2
CAD
TCD0103-025G
TCD0103-100G
4-Nitro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
4-Nitro-o-phenylenediamine
CAS Number: 99-56-9
MDL Number: MFCD00007724
Molecular Formula: C6H7N3O2
Molecular Weight: 153.14
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 201
Other Product Names / Synonyms: 1,2-Diamino-4-nitrobenzene
Formula:
C₆H₇N₃O₂ Melting Pt: 201 °C |
MDL Number:
MFCD00007724 CAS Number: 99-56-9 EINECS: 202-766-3 Merck Index: 12,06720 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...