1,8-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
D0102-25G
D0102-500G
D0102-100G
TCD0102-025GEA
175.38
CAD
TCD0102-025G
TCD0102-500G
TCD0102-100G
1,8-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
1,8-Diaminonaphthalene
CAS Number: 479-27-6
MDL Number: MFCD00004033
Molecular Formula: C10H10N2
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 65
Other Product Names / Synonyms: 1,8-Naphthalenediamine
Formula:
C₁₀H₁₀N₂ Boiling Pt: 205 °C Melting Pt: 65 °C Density: 1.18 g/cm³ (20 °C) Flash Pt: 195 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004033 CAS Number: 479-27-6 EINECS: 207-529-8 UN: 3259 ADR: 8,III Merck Index: 12,06459 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...