N,N-Diethyl-m-toluamide ≥98.0%

Supplier: TCI America
Warning

D0097-500ML D0097-25ML
TCD0097-500MLEA 209.38 CAD
TCD0097-500ML TCD0097-025ML
N,N-Diethyl-m-toluamide ≥98.0%
N,N-Diethyl-m-toluamide

CAS Number: 134-62-3
MDL Number: MFCD00009046
Molecular Formula: C12H17NO
Molecular Weight: 191.27
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 140
Specific Gravity (20/20): 1.00

Formula: C₁₂H₁₇NO
Boiling Pt: 160 °C
Melting Pt: –45 °C
Density: 1.00 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00009046
CAS Number: 134-62-3
EINECS: 205-149-7
Merck Index: 13,02876
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d1f286102c19a2f0c1d30c77b41c588e5ffb0ab&vid=2d1f286102c19a2f0c1d30c77b41c588e5ffb0ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR