1,12-Diaminododecane ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: N12N, Dodecamethylendiamine, 1,12-Dodecanediamine

D0091-500G D0091-25G D0091-100G
TCD0091-500GEA 1116.63 CAD
TCD0091-500G TCD0091-025G TCD0091-100G
1,12-Diaminododecane ≥98.0% (by GC, titration analysis)
1,12-Diaminododecane

CAS Number: 2783-17-7
MDL Number: MFCD00008155
Molecular Formula: C12H28N2
Molecular Weight: 200.37
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 70
Flash Point (°C): 155


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Dodecamethylenediamine, 1,12-Dodecanediamine

Formula: C₁₂H₂₈N₂
MW: 200.37 g/mol
Boiling Pt: 145 °C
Melting Pt: 70 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00008155
CAS Number: 2783-17-7
EINECS: 220-489-6
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d62b49709ff6862a275b92f1d650ed376d90029&vid=3d62b49709ff6862a275b92f1d650ed376d90029&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR