Dapsone ≥97.0%

Supplier: TCI America
Warning

Synonyms: 4-Aminophenyl sulfone, 1-Amino-4-((4-aminophenyl)sulfonyl)benzene, 4,4'-sulfonyldianiline, 4,4'-Sulfonyldianiline, 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Diamino diphenyl sulfone, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone, 4-[(4-Aminophenyl)sulfonyl]aniline

D0089-25G D0089-500G D0089-100G
TCD0089-025GEA 41.81 CAD
TCD0089-025G TCD0089-500G TCD0089-100G
Dapsone ≥97.0%
Dapsone

CAS Number: 80-08-0
MDL Number: MFCD00007887
Molecular Formula: C12H12N2O2S
Molecular Weight: 248.30
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 177


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials, Diphenyl Sulfones (for High-Performance Polymer Research), Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: 4,4'-Diaminodiphenyl Sulfone, 4,4'-Sulfonyldianiline

Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00007887
CAS Number: 80-08-0
EINECS: 201-248-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9e07da64f921c4e066d4b7699b72ab740df1959&vid=d9e07da64f921c4e066d4b7699b72ab740df1959&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR