1,4-Diaminobutane dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Butanediamine dihydrochloride, Tetramethylenediammonium dichloride, Tetramethylendiamine dihydrochloride, Putrescine dihydrochloride

D0081-500G D0081-25G
TCD0081-500GEA 636.74 CAD
TCD0081-500G TCD0081-025G
1,4-Diaminobutane dihydrochloride ≥98.0%
1,4-Diaminobutane dihydrochloride

CAS Number: 333-93-7
MDL Number: MFCD00012526
Molecular Formula: C4H12N2
Molecular Weight: 161.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Monomers, Bioscience, Regenerative Medicine Research Reagents, Polyamines
Other Product Names / Synonyms: 1,4-Butanediamine Dihydrochloride, Putrescine Dihydrochloride, Tetramethylenediamine Dihydrochloride

Formula: C₄H₁₄Cl₂N₂
MW: 161.07 g/mol
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00012526
CAS Number: 333-93-7
EINECS: 206-375-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db36cdb25dff162482d60fe6d38c6d334f5bed53&vid=db36cdb25dff162482d60fe6d38c6d334f5bed53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR