1,4-Diamino-9,10-anthraquinone ≥97.0%

Supplier: TCI America

Synonyms: 1,4-Diaminoanthraquinone

D0075-25G D0075-1G
TCD0075-25GEA 851.62 CAD
TCD0075-25G TCD0075-001G
1,4-Diamino-9,10-anthraquinone ≥97.0%
1,4-Diamino-9,10-anthraquinone

CAS Number: 128-95-0
MDL Number: MFCD00001224
Molecular Formula: C14H10N2O2
Molecular Weight: 238.25
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 270
Flash Point (°C): 340

Formula: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
Melting Pt: 270 °C
Flash Pt: 340 °C
MDL Number: MFCD00001224
CAS Number: 128-95-0
EINECS: 204-922-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=476917133669c007add170226d1bc35d7cb8cf71&vid=476917133669c007add170226d1bc35d7cb8cf71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR