2'-Deoxyuridine ≥99.0% (by HPLC)

Supplier: TCI America

D0060-25G D0060-1G D0060-5G
TCD0060-25GEA 788.42 CAD
TCD0060-25G TCD0060-001G TCD0060-005G
2'-Deoxyuridine ≥99.0% (by HPLC)
2'-Deoxyuridine

CAS Number: 951-78-0
MDL Number: MFCD00006527
Molecular Formula: C9H12N2O5
Molecular Weight: 228.20
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: 52 deg (C=1, 1mol/L NaOH sol.)

Formula: C₉H₁₂N₂O₅
MW: 228.20 g/mol
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00006527
CAS Number: 951-78-0
EINECS: 213-455-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4286e77149b42e0bb559960e74940a0d86acf1a6&vid=4286e77149b42e0bb559960e74940a0d86acf1a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR