2-Deoxy-D(-)-ribose 98,0

Supplier: TCI America

Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal

D0059-25G D0059-5G
TCD0059-025GEA 157.08 CAD
TCD0059-025G TCD0059-5G
2-Deoxy-D(-)-ribose 98,0
2-Deoxy-D(-)-ribose

CAS Number: 533-67-5
MDL Number: MFCD00135904
Molecular Formula: C5H10O4
Molecular Weight: 134.13
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: -56 deg (C=1, H2O)

Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 292
Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00135904
CAS Number: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d706a5735ac783ad33a055c386e15b3d96d24fe&vid=9d706a5735ac783ad33a055c386e15b3d96d24fe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR