2'-Deoxyguanosine hydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)

D0052-5G D0052-25G D0052-1G
TCD0052-005GEA 241.08 CAD
TCD0052-005G TCD0052-25G TCD0052-001G
2'-Deoxyguanosine hydrate ≥98.0% (by titrimetric analysis)
2'-Deoxyguanosine hydrate

CAS Number: 961-07-9
MDL Number: MFCD00080300
Molecular Formula: C10H13N5O4
Molecular Weight: 267.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -42 deg (C=0.2, 1mol/L NaOH sol.)

Formula: C₁₀H₁₃N₅O₄·H₂O
MW: 267.24 g/mol (anhy)
MDL Number: MFCD00150760
CAS Number: 961-07-9
EINECS: 213-505-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e97eb5f40600988ac08f56b0416c82256cb6114e&vid=6eaa23cd707fce4d8433d8f08d756797ca910cf3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR