2-Deoxy-D-glucose ≥97.0%

Supplier: TCI America

Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

D0051-1G D0051-5G D0051-25G
TCD0051-001GEA 77.74 CAD
TCD0051-001G TCD0051-5G TCD0051-25G
2-Deoxy-D-glucose ≥97.0%
2-Deoxy-D-glucose

CAS Number: 154-17-6
MDL Number: MFCD00151328
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 46.5 deg (C=1, H2O)

Formula: C₆H₁₂O₅
MW: 164.16 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00151328
CAS Number: 154-17-6
EINECS: 205-823-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87caa302445f6f20523c3de710090f7bc9f797cf&vid=87caa302445f6f20523c3de710090f7bc9f797cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR