2'-Deoxycytidine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D0048-25G
D0048-5G
TCD0048-025GEA
619.83
CAD
TCD0048-025G
TCD0048-005G
2'-Deoxycytidine hydrochloride ≥98.0% (by titrimetric analysis)
2'-Deoxycytidine hydrochloride
CAS Number: 3992-42-5
MDL Number: MFCD00012840
Molecular Formula: C9H13N3O4
Molecular Weight: 263.68
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Flash Point (°C): 162
Specific rotation [a]20/D: 57 deg (C=1, H2O)
Formula:
C₉H₁₄ClN₃O₄ Flash Pt: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012840 CAS Number: 3992-42-5 EINECS: 223-639-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...