Dehydroacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione, 3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

D0039-500G D0039-25G
TCD0039-500GEA 166.55 CAD
TCD0039-500G TCD0039-025G
Dehydroacetic acid ≥98.0% (by GC, titration analysis)
Dehydracetic acid

CAS Number: 520-45-6
MDL Number: MFCD00066709
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 270
Melting point (°C): 111
Flash Point (°C): 157

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 270 °C
Melting Pt: 111 °C
Flash Pt: 157
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46a4c6456e7d34c97ae496cdb158044b1d9004d7&vid=46a4c6456e7d34c97ae496cdb158044b1d9004d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR