1-Phenyl-1-decanone ≥98.0%

Supplier: TCI America

D0026-500G D0026-25G
TCD0026-500GEA 665.18 CAD
TCD0026-500G TCD0026-025G
1-Phenyl-1-decanone ≥98.0%
1-Phenyl-1-decanone

CAS Number: 6048-82-4
MDL Number: MFCD00009582
Molecular Formula: C16H24O
Molecular Weight: 232.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 35
Flash Point (°C): 110


Other Product Names / Synonyms: Nonyl Phenyl Ketone

Formula: C₆H₅C(O)(CH₂)₈CH₃
Boiling Pt: 187 °C
Melting Pt: 35 °C
Flash Pt: 110 °C
MDL Number: MFCD00009582
CAS Number: 6048-82-4
EINECS: 227-946-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5444823eec7734a2d12e7d59cb303fadaa31e7c&vid=958f68737ca796321bb540fa6b05c8b4903f5311&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR