Methyl decanoate ≥98.0%
Supplier: TCI America
D0023-25ML
D0023-500ML
TCD0023-025MLEA
56.55
CAD
TCD0023-025ML
TCD0023-500ML
Methyl decanoate ≥98.0%
Methyl decanoate
CAS Number: 110-42-9
MDL Number: MFCD00009580
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Melting point (°C): -18
Flash Point (°C): 107
Specific Gravity (20/20): 0.87
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Decanoic Acid Methyl Ester
Formula:
C₁₁H₂₂O₂ Boiling Pt: 229 °C Melting Pt: –18 °C Density: 0.87 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00009580 CAS Number: 110-42-9 EINECS: 203-766-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...